Product Details

SNP ID
rs113135397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36380634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGGGAGCCCAACACCGTTCCCG[C/T]GCGGCCACGATGATCCCTCCGAGCA
Phenotype
MIM: 602317
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STAC PubMed Links

Gene Details

Gene
STAC
Gene Name
SH3 and cysteine rich domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292049.1 180 UTR 5 NP_001278978.1
NM_003149.2 180 UTR 5 NP_003140.1
XM_011534036.2 180 Intron XP_011532338.1
XM_011534037.2 180 UTR 5 XP_011532339.1
XM_011534038.2 180 Intron XP_011532340.1
XM_017007083.1 180 Intron XP_016862572.1
XM_017007084.1 180 Intron XP_016862573.1

View Full Product Details