Product Details

SNP ID
rs113903336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39183987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCTGTCAGGTCACTGCTGAAC[C/T]CAGCTGGGCTGTGCAGGAACTGCCT
Phenotype
MIM: 609777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XIRP1 PubMed Links

Gene Details

Gene
XIRP1
Gene Name
xin actin binding repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198621.2 5263 UTR 3 NP_001185550.1
NM_194293.2 5263 Missense Mutation NP_919269.2
XM_005264909.3 5263 Missense Mutation XP_005264966.1

View Full Product Details