Product Details

SNP ID
rs113502502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39332382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAACAGTGACCGACTACTACTA[C/T]CCTGATATCTTCTCAAGCCCCTGTG
Phenotype
MIM: 601834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR8 PubMed Links

Gene Details

Gene
CCR8
Gene Name
C-C motif chemokine receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005201.3 189 Silent Mutation TAC,TAT Y17Y NP_005192.1

View Full Product Details