Product Details

SNP ID
rs114247597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:43289192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGCGGCAGTGAACAACCTGAC[A/G]TGGAAATGCTTCACAGAGGAGGTGA
Phenotype
MIM: 612760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNRK PubMed Links

Gene Details

Gene
SNRK
Gene Name
SNF related kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100594.1 Intron NP_001094064.1
NM_017719.4 Intron NP_060189.3
XM_005265245.3 Intron XP_005265302.1
XM_011533861.2 Intron XP_011532163.1

View Full Product Details