Product Details
- SNP ID
-
rs116103451
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44755596 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAGCTCCTTTGCTTCCCCTGTCT[C/T]TGAACAAATTCCTACGGCTTATTAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1143
PubMed Links
Gene Details
- Gene
- KIAA1143
- Gene Name
- KIAA1143
- Gene
- KIF15
- Gene Name
- kinesin family member 15
There are no transcripts associated with this gene.
View Full Product Details