Product Details

SNP ID
rs116892735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44862905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTAGTATCCCATTGGGTGTAGT[C/T]CATGGGGTAAGTGGTCGAAAGTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF15 PubMed Links

Gene Details

Gene
KIF15
Gene Name
kinesin family member 15
There are no transcripts associated with this gene.

Gene
MIR564
Gene Name
microRNA 564
There are no transcripts associated with this gene.

Gene
TMEM42
Gene Name
transmembrane protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144638.2 Intron NP_653239.1

View Full Product Details