Product Details

SNP ID
rs112023020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50269313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAGTGGGGCTGGGGAGTGCC[A/G]CCCCCAGCCCCCCACGCCTTCGGCT
Phenotype
MIM: 139360 MIM: 601281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAI2 PubMed Links

Gene Details

Gene
GNAI2
Gene Name
G protein subunit alpha i2
There are no transcripts associated with this gene.

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
There are no transcripts associated with this gene.

Gene
MIR6872
Gene Name
microRNA 6872
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005914.2 365 Missense Mutation ACC,GCC T25A NP_001005914.1
NM_001290060.1 365 Missense Mutation ACC,GCC T25A NP_001276989.1
NM_001290061.1 365 Missense Mutation ACC,GCC T25A NP_001276990.1
NM_001290062.1 365 Intron NP_001276991.1
NM_001290063.1 365 Intron NP_001276992.1
NM_004636.3 365 Missense Mutation ACC,GCC T25A NP_004627.1
Gene
SEMA3B-AS1
Gene Name
SEMA3B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details