Product Details

SNP ID
rs115745575
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52719292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTGGAGATGTAAACTAAACTA[C/T]AGGGTGGGCTAGTTTGCAGAGACGA
Phenotype
MIM: 601959
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NEK4 PubMed Links

Gene Details

Gene
NEK4
Gene Name
NIMA related kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193533.1 Intron NP_001180462.1
NM_003157.4 Intron NP_003148.2
XM_006713310.2 Intron XP_006713373.1
XM_011534039.2 Intron XP_011532341.1
XM_011534040.2 Intron XP_011532342.1
XM_017007085.1 Intron XP_016862574.1
XM_017007086.1 Intron XP_016862575.1

View Full Product Details