Product Details

SNP ID
rs116121608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53287414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATGCTTCACAGTGAAACCTCC[A/G]TTATCACTGAGAATGTCACTTGGAA
Phenotype
MIM: 607010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCP1A PubMed Links

Gene Details

Gene
DCP1A
Gene Name
decapping mRNA 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290204.1 1911 UTR 3 NP_001277133.1
NM_001290205.1 1911 UTR 3 NP_001277134.1
NM_001290206.1 1911 UTR 3 NP_001277135.1
NM_001290207.1 1911 UTR 3 NP_001277136.1
NM_018403.6 1911 UTR 3 NP_060873.4

View Full Product Details