Product Details

SNP ID
rs114294662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58306283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACGCTTCGTGGGGCGGGACGA[A/G]GAGAAGCCAAACGTAAAGACACCAG
Phenotype
MIM: 606112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPP14 PubMed Links

Gene Details

Gene
RPP14
Gene Name
ribonuclease P/MRP subunit p14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098783.2 39 UTR 5 NP_001092253.1
NM_007042.4 39 UTR 5 NP_008973.1
XM_011533313.2 39 Intron XP_011531615.1
XM_017005641.1 39 UTR 5 XP_016861130.1

View Full Product Details