Product Details

SNP ID
rs114051832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69934649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACTAATCCTACATTCTGTCCTG[A/C]AATTGGAATAATACCATTTAAAAAT
Phenotype
MIM: 115450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSN1S1 PubMed Links

Gene Details

Gene
CSN1S1
Gene Name
casein alpha s1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025104.1 Intron NP_001020275.1
NM_001890.1 Intron NP_001881.1
XM_006714089.2 Intron XP_006714152.1
XM_006714090.2 Intron XP_006714153.1
XM_006714091.2 Intron XP_006714154.1

View Full Product Details