Product Details

SNP ID
rs113120280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70817542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTAAGTATATGAAAAGGTGCTC[C/G]ACATCACTATGGGATTGGGAATTAA
Phenotype
MIM: 604851 MIM: 611194
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRSF1 PubMed Links

Gene Details

Gene
GRSF1
Gene Name
G-rich RNA sequence binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098477.1 4707 UTR 3 NP_001091947.1
NM_002092.3 4707 UTR 3 NP_002083.3
XM_005265681.3 4707 Intron XP_005265738.1
XM_011531897.2 4707 Intron XP_011530199.1
Gene
RUFY3
Gene Name
RUN and FYVE domain containing 3
There are no transcripts associated with this gene.

View Full Product Details