Product Details

SNP ID
rs117987138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:71743295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGAAATGAAACACATAGCCAA[A/C]CAAAGTTGGCTTAATGGTTTTAGGA
Phenotype
MIM: 139200
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GC PubMed Links

Gene Details

Gene
GC
Gene Name
GC, vitamin D binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000583.3 Intron NP_000574.2
NM_001204306.1 Intron NP_001191235.1
NM_001204307.1 Intron NP_001191236.1
XM_006714177.2 Intron XP_006714240.1

View Full Product Details