Product Details
- SNP ID
-
rs111283987
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73981208 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACACGTAGGCAGCTAGTAGCTAA[A/C]TCTCCAAAAGTTTCTTAATTATTTT
- Phenotype
-
MIM: 173460
MIM: 121010
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs352006] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PF4
- Gene Name
- platelet factor 4
- Gene
- PPBP
- Gene Name
- pro-platelet basic protein
There are no transcripts associated with this gene.
View Full Product Details