Product Details

SNP ID
rs116796864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73981881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCACCGCTGGCGAAGGCGACCA[C/T]AAGTGGCAGGAGCAGCAACCCCAGG
Phenotype
MIM: 173460 MIM: 121010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PF4 PubMed Links

Gene Details

Gene
PF4
Gene Name
platelet factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002619.3 244 Missense Mutation ATG,GTG M25V NP_002610.1
XM_005265696.3 244 Intron XP_005265753.1
Gene
PPBP
Gene Name
pro-platelet basic protein
There are no transcripts associated with this gene.

View Full Product Details