Product Details

SNP ID
rs114978073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67565412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAGAAAATATTTCCTCCTTCTC[A/G]TACATATTACCATTGAAGGAGATCT
Phenotype
MIM: 604298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAP1 PubMed Links

Gene Details

Gene
STAP1
Gene Name
signal transducing adaptor family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317769.1 Intron NP_001304698.1
NM_012108.3 Intron NP_036240.1
XM_017008018.1 Intron XP_016863507.1

View Full Product Details