Product Details

SNP ID
rs114812738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67617694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCAAATTGGAGTAATATTGATG[C/T]TAATCTAGTTTTTTATTTAACAAAG
Phenotype
MIM: 611361
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBA6 PubMed Links

Gene Details

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 4514 UTR 3 NP_060697.4
XM_017008359.1 4514 Intron XP_016863848.1

View Full Product Details