Product Details

SNP ID
rs116326015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:53996486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGAAAGAACCAAAACACTGTT[A/C]ATGTCTATCCACTCATGTGTGAAAA
Phenotype
MIM: 604332
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHIC2 PubMed Links

Gene Details

Gene
CHIC2
Gene Name
cysteine rich hydrophobic domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012110.3 14371 Intron NP_036242.1
XM_006714037.3 14371 UTR 3 XP_006714100.1
XM_011534382.2 14371 Intron XP_011532684.1
Gene
RPL21P44
Gene Name
ribosomal protein L21 pseudogene 44
There are no transcripts associated with this gene.

View Full Product Details