Product Details
- SNP ID
-
rs115581241
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:55398328 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGTTACTGGGAAGTGAAGATTC[C/T]GGGAGCCGATTTAATCCTCACAACA
- Phenotype
-
MIM: 614726
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM165
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142514414] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM165
- Gene Name
- transmembrane protein 165
View Full Product Details