Product Details

SNP ID
rs112496764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56469687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACAAAGATGACGTAACTGCCCT[G/T]CATTGTAAAGTGGTATGCCTTATCC
Phenotype
MIM: 172439 MIM: 602122
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAICS PubMed Links

Gene Details

Gene
PAICS
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
There are no transcripts associated with this gene.

Gene
SRP72
Gene Name
signal recognition particle 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267722.1 184 Silent Mutation CTG,CTT L48L NP_001254651.1
NM_006947.3 184 Silent Mutation CTG,CTT L48L NP_008878.3
XM_005265765.4 184 Silent Mutation CTG,CTT L163L XP_005265822.3

View Full Product Details