Product Details

SNP ID
rs112351543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56517171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGAAATGAGGAACAACTCAAAC[A/G]TCCACTGATGTTTAAACTATGTGTG
Phenotype
MIM: 612405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL9 PubMed Links

Gene Details

Gene
ARL9
Gene Name
ADP ribosylation factor like GTPase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206919.1 Intron NP_996802.1
XM_005265719.3 Intron XP_005265776.1
XM_005265720.3 Intron XP_005265777.1
XM_006714036.3 Intron XP_006714099.1

View Full Product Details