Product Details

SNP ID
rs114568376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176553606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAGGTAGGTTCCAGGTTACAG[C/T]CTCAGAGGGATGCCTGGGCTGGACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171976.1 Intron NP_001165447.1
NM_017675.4 Intron NP_060145.3

View Full Product Details