Product Details
- SNP ID
-
rs115622094
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178865109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATTGGAACCTTTAATGTGAGCCC[A/G]ATATGGTTTTAAGTGCATTGTGCAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF354B
PubMed Links
Gene Details
- Gene
- ZNF354B
- Gene Name
- zinc finger protein 354B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_058230.2 |
|
Intron |
|
|
NP_478137.1 |
View Full Product Details