Product Details

SNP ID
rs117600221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178980286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTAAATTCTACTCCCAGGGACA[G/T]AAAACTCCATAACTTAAACCAAGCT
Phenotype
MIM: 604096
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRM6 PubMed Links

Gene Details

Gene
GRM6
Gene Name
glutamate metabotropic receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000843.3 4184 UTR 3 NP_000834.2

View Full Product Details