Product Details

SNP ID
rs112372284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181224497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGTTGAGGAAGAGGATCTAGA[C/T]CCCGTCACCCCACTGCCCCCGCCTC
Phenotype
MIM: 610530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4638 PubMed Links

Gene Details

Gene
MIR4638
Gene Name
microRNA 4638
There are no transcripts associated with this gene.

Gene
TRIM41
Gene Name
tripartite motif containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033549.4 1233 Silent Mutation GAC,GAT D166D NP_291027.3
NM_201627.2 1233 Silent Mutation GAC,GAT D166D NP_963921.1
XM_006714929.2 1233 Silent Mutation GAC,GAT D166D XP_006714992.1
XM_006714930.2 1233 Silent Mutation GAC,GAT D166D XP_006714993.1
XM_006714931.3 1233 Silent Mutation GAC,GAT D166D XP_006714994.1
XM_006714932.3 1233 Silent Mutation GAC,GAT D166D XP_006714995.1

View Full Product Details