Product Details

SNP ID
rs116159251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:158699332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACATGTGCTTTATTAAAAATT[A/G]TAATAAATCAGCATGGGTATGTGTC
Phenotype
MIM: 164343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EBF1 PubMed Links

Gene Details

Gene
EBF1
Gene Name
early B-cell factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290360.2 Intron NP_001277289.1
NM_001324101.1 Intron NP_001311030.1
NM_001324103.1 Intron NP_001311032.1
NM_001324106.1 Intron NP_001311035.1
NM_001324107.1 Intron NP_001311036.1
NM_001324108.1 Intron NP_001311037.1
NM_001324109.1 Intron NP_001311038.1
NM_001324111.1 Intron NP_001311040.1
NM_024007.4 Intron NP_076870.1
NM_182708.2 Intron NP_874367.1
XM_017009192.1 Intron XP_016864681.1
XM_017009193.1 Intron XP_016864682.1
XM_017009194.1 Intron XP_016864683.1
XM_017009195.1 Intron XP_016864684.1
XM_017009196.1 Intron XP_016864685.1
XM_017009197.1 Intron XP_016864686.1
XM_017009198.1 Intron XP_016864687.1
XM_017009199.1 Intron XP_016864688.1
XM_017009200.1 Intron XP_016864689.1
XM_017009201.1 Intron XP_016864690.1
XM_017009202.1 Intron XP_016864691.1
XM_017009203.1 Intron XP_016864692.1
XM_017009204.1 Intron XP_016864693.1

View Full Product Details