Product Details

SNP ID
rs117375974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160394047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAACTATCATCAACAAAATCGA[C/T]ATTAAAAAGAAATGGATCCAATATC
Phenotype
MIM: 605974 MIM: 615253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLU7 PubMed Links

Gene Details

Gene
SLU7
Gene Name
SLU7 homolog, splicing factor
There are no transcripts associated with this gene.

Gene
ZBED8
Gene Name
zinc finger BED-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303251.1 2026 Missense Mutation ATC,GTC I482V NP_001290180.1
NM_022090.4 2026 Missense Mutation ATC,GTC I482V NP_071373.2

View Full Product Details