Product Details

SNP ID
rs118104999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136759670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGTTTATTGAATAGAAAGAGA[C/T]GTCCCACAAGGCATCCCACAGGCGG
Phenotype
MIM: 612902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCN15 PubMed Links

Gene Details

Gene
LCN15
Gene Name
lipocalin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203347.1 654 UTR 3 NP_976222.1
XM_011518672.1 654 UTR 3 XP_011516974.1
Gene
LCN8
Gene Name
lipocalin 8
There are no transcripts associated with this gene.

Gene
LOC100128593
Gene Name
uncharacterized LOC100128593
There are no transcripts associated with this gene.

View Full Product Details