Product Details

SNP ID
rs112492960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137220538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCTCGGGCTCAGCCCCCCACTG[A/T]CCCCCGGATGGGGCCGTCAGCGCCT
Phenotype
MIM: 606073
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYSRT1 PubMed Links

Gene Details

Gene
CYSRT1
Gene Name
cysteine rich tail 1
There are no transcripts associated with this gene.

Gene
NDOR1
Gene Name
NADPH dependent diflavin oxidoreductase 1
There are no transcripts associated with this gene.

Gene
RNF208
Gene Name
ring finger protein 208
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031297.4 786 Silent Mutation GGA,GGT G225G NP_112587.2
Gene
RNF224
Gene Name
ring finger protein 224
There are no transcripts associated with this gene.

View Full Product Details