Product Details

SNP ID
rs116305810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137244256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTGTGTTCCAGGCAGCTTCTC[A/G]CCCAGGTCACAGTGGGGGTTTCTAA
Phenotype
MIM: 609826 MIM: 602660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001710.2 1747 Silent Mutation GGA,GGG G260G NP_001001710.1
XM_006717108.2 1747 Silent Mutation GGA,GGG G244G XP_006717171.2
XM_011518694.1 1747 Silent Mutation GGA,GGG G295G XP_011516996.1
XM_011518695.1 1747 Silent Mutation GGA,GGG G257G XP_011516997.1
XM_011518696.2 1747 Intron XP_011516998.1
XM_011518697.1 1747 UTR 3 XP_011516999.1
XM_017014718.1 1747 Silent Mutation GGA,GGG G173G XP_016870207.1
Gene
SLC34A3
Gene Name
solute carrier family 34 member 3
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
There are no transcripts associated with this gene.

Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006088.5 1747 Intron NP_006079.1

View Full Product Details