Product Details

SNP ID
rs116185717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132496619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCGTCTGCACATGGCTGTGAC[A/G]TTGCTGCTTCCCTGCGGCTGCGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBRSL1 PubMed Links

Gene Details

Gene
FBRSL1
Gene Name
fibrosin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142641.1 Intron NP_001136113.1
XM_005266171.3 Intron XP_005266228.1
XM_005266173.3 Intron XP_005266230.1
XM_005266175.3 Intron XP_005266232.1
XM_005266176.3 Intron XP_005266233.1
XM_005266177.3 Intron XP_005266234.1
XM_005266181.3 Intron XP_005266238.1
XM_011534803.2 Intron XP_011533105.1
XM_011534804.2 Intron XP_011533106.1
XM_011534805.2 Intron XP_011533107.1
XM_011534806.2 Intron XP_011533108.1
XM_011534807.2 Intron XP_011533109.1
XM_011534808.2 Intron XP_011533110.1
XM_011534809.2 Intron XP_011533111.1
XM_011534810.2 Intron XP_011533112.1
XM_011534812.2 Intron XP_011533114.1
XM_011534816.2 Intron XP_011533118.1

View Full Product Details