Product Details

SNP ID
rs113568898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132773170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCTGTGGGTCGCCGCGTGGGC[C/T]GGCGTGACCCCCCGGGGGCACAGGG
Phenotype
MIM: 602581
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGA3 PubMed Links

Gene Details

Gene
GOLGA3
Gene Name
golgin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172557.1 2596 Intron NP_001166028.1
NM_005895.3 2596 Missense Mutation AGC,GGC S1478G NP_005886.2
XM_005266164.4 2596 Missense Mutation AGC,GGC S1438G XP_005266221.1
XM_005266167.4 2596 Missense Mutation AGC,GGC S1287G XP_005266224.1
XM_006719736.3 2596 Missense Mutation AGC,GGC S1478G XP_006719799.1
XM_006719737.3 2596 Missense Mutation AGC,GGC S1478G XP_006719800.1
XM_011534793.2 2596 Missense Mutation AGC,GGC S1387G XP_011533095.1
XM_011534794.2 2596 Missense Mutation AGC,GGC S830G XP_011533096.1

View Full Product Details