Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135955.1 | 3515 | Intron | NP_001129427.1 | ||
NM_001135956.1 | 3515 | Intron | NP_001129428.1 | ||
NM_001135957.1 | 3515 | Intron | NP_001129429.1 | ||
NM_001135958.1 | 3515 | Intron | NP_001129430.1 | ||
NM_003306.1 | 3515 | Intron | NP_003297.1 | ||
NM_016179.2 | 3515 | Intron | NP_057263.1 | ||
XM_011535206.1 | 3515 | UTR 3 | XP_011533508.1 | ||
XM_017020723.1 | 3515 | UTR 3 | XP_016876212.1 | ||
XM_017020724.1 | 3515 | Intron | XP_016876213.1 |