Product Details

SNP ID
rs114600063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:37637202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTTGAATATTTCTCTCAAGTG[A/G]TCCTGCAGCCTGTTGACGAGCAACT
Phenotype
MIM: 603651
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPC4 PubMed Links

Gene Details

Gene
TRPC4
Gene Name
transient receptor potential cation channel subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135955.1 3431 Missense Mutation NP_001129427.1
NM_001135956.1 3431 Missense Mutation NP_001129428.1
NM_001135957.1 3431 Missense Mutation NP_001129429.1
NM_001135958.1 3431 Missense Mutation NP_001129430.1
NM_003306.1 3431 Missense Mutation NP_003297.1
NM_016179.2 3431 Missense Mutation NP_057263.1
XM_011535206.1 3431 Missense Mutation XP_011533508.1
XM_017020723.1 3431 Missense Mutation XP_016876212.1
XM_017020724.1 3431 Missense Mutation XP_016876213.1

View Full Product Details