Product Details

SNP ID
rs116617950
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40556620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTGCTTTCCAGACAGACCAGA[C/T]GCCTTTCCCTGGACTTCACTGTTCT
Phenotype
MIM: 136533
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FOXO1 PubMed Links

Gene Details

Gene
FOXO1
Gene Name
forkhead box O1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002015.3 4782 UTR 3 NP_002006.2
XM_011535008.2 4782 UTR 3 XP_011533310.1
XM_011535010.2 4782 UTR 3 XP_011533312.1

View Full Product Details