Product Details

SNP ID
rs113753087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45471123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGCCTTACAGCAGGATTCTTTG[G/T]CTGGGCATGGTGGCTCACCCCTGTA
Phenotype
MIM: 606975
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COG3 PubMed Links

Gene Details

Gene
COG3
Gene Name
component of oligomeric golgi complex 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031431.3 Intron NP_113619.2
XM_011535266.1 Intron XP_011533568.1
XM_011535267.2 Intron XP_011533569.1
XM_017020790.1 Intron XP_016876279.1

View Full Product Details