Product Details

SNP ID
rs112950191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50015976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTAACTTTTTCCCTCCTCAGAT[A/G]ACCTTACTTCCACTGCCTCCACAAA
Phenotype
MIM: 605766 MIM: 607947 MIM: 605661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLEU2 PubMed Links

Gene Details

Gene
DLEU2
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
KCNRG
Gene Name
potassium channel regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173605.1 723 Missense Mutation ATA,ATG I161M NP_775876.1
NM_199464.2 723 Missense Mutation ATA,ATG I161M NP_955751.1
Gene
TRIM13
Gene Name
tripartite motif containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007278.2 723 UTR 3 NP_001007279.1
NM_005798.4 723 UTR 3 NP_005789.2
NM_052811.3 723 UTR 3 NP_434698.1
NM_213590.2 723 UTR 3 NP_998755.1

View Full Product Details