Product Details

SNP ID
rs114298305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51815572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACAGCTCAGCAGTGTGTAAATA[C/T]ACAGAAATGCTATGCACGTGGCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00282 PubMed Links

Gene Details

Gene
LINC00282
Gene Name
long intergenic non-protein coding RNA 282
There are no transcripts associated with this gene.

View Full Product Details