Product Details

SNP ID
rs113698407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:61411518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTCCTCTATATTAATCTCTG[A/G]TTCTTTTCTTAAAAGACTCTCAATG
Phenotype
MIM: 614449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH20 PubMed Links

Gene Details

Gene
PCDH20
Gene Name
protocadherin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022843.3 4381 Missense Mutation CCA,TCA P861S NP_073754.2
XM_017020708.1 4381 Missense Mutation CCA,TCA P861S XP_016876197.1

View Full Product Details