Product Details

SNP ID
rs113336140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23570583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGAGTCTGTGGACTAGGAGG[C/T]GGGAGGGTAACTACCTGCTGAAAGT
Phenotype
MIM: 606122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF19 PubMed Links

Gene Details

Gene
TNFRSF19
Gene Name
TNF receptor superfamily member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204458.1 214 Intron NP_001191387.1
NM_001204459.1 214 UTR 5 NP_001191388.1
NM_018647.3 214 Intron NP_061117.2
NM_148957.3 214 UTR 5 NP_683760.1
XM_005266445.2 214 UTR 5 XP_005266502.1
XM_005266446.1 214 UTR 5 XP_005266503.1
XM_011535146.1 214 Intron XP_011533448.1
XM_017020651.1 214 UTR 5 XP_016876140.1
XM_017020652.1 214 Intron XP_016876141.1

View Full Product Details