Product Details
- SNP ID
-
rs114118520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:76884177 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCCTCCCTCCCCACCCCACCCC[A/G]CCCACCAAGTCTTACAGCTAAAGGA
- Phenotype
-
MIM: 610521
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201622249] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD12
- Gene Name
- potassium channel tetramerization domain containing 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138444.3 |
2229 |
UTR 3 |
|
|
NP_612453.1 |
View Full Product Details