Product Details

SNP ID
rs117661532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:79410544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTTAAAAACAGTCAGCATTTC[C/T]GGATCAAAAAGAGATTCTGTATTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM26 PubMed Links

Gene Details

Gene
RBM26
Gene Name
RNA binding motif protein 26
There are no transcripts associated with this gene.

Gene
RBM26-AS1
Gene Name
RBM26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details