Product Details

SNP ID
rs117629222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76357836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGCTGAGTCAGAATTCCGTC[C/T]GCGTCCATCCCTGTGCGTCCTGTAT
Phenotype
MIM: 615114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF516 PubMed Links

Gene Details

Gene
ZNF516
Gene Name
zinc finger protein 516
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014643.3 8472 UTR 3 NP_055458.1
XM_011526269.1 8472 Intron XP_011524571.1
XM_011526270.2 8472 Intron XP_011524572.1
XM_011526271.2 8472 Intron XP_011524573.1
XM_011526272.2 8472 Intron XP_011524574.1
XM_011526273.2 8472 Intron XP_011524575.1
XM_011526274.2 8472 Intron XP_011524576.1
XM_011526275.2 8472 Intron XP_011524577.1
XM_011526276.1 8472 Intron XP_011524578.1
XM_011526277.2 8472 Intron XP_011524579.1
XM_017026097.1 8472 Intron XP_016881586.1

View Full Product Details