Product Details

SNP ID
rs116197317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76358541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCAACCACGGCAAACAAAGGC[A/G]TCCTCCTCACTTGAAGTCCTGGCCT
Phenotype
MIM: 615114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF516 PubMed Links

Gene Details

Gene
ZNF516
Gene Name
zinc finger protein 516
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014643.3 7718 UTR 3 NP_055458.1
XM_011526269.1 7718 UTR 3 XP_011524571.1
XM_011526270.2 7718 UTR 3 XP_011524572.1
XM_011526271.2 7718 UTR 3 XP_011524573.1
XM_011526272.2 7718 UTR 3 XP_011524574.1
XM_011526273.2 7718 UTR 3 XP_011524575.1
XM_011526274.2 7718 UTR 3 XP_011524576.1
XM_011526275.2 7718 UTR 3 XP_011524577.1
XM_011526276.1 7718 UTR 3 XP_011524578.1
XM_011526277.2 7718 Intron XP_011524579.1
XM_017026097.1 7718 UTR 3 XP_016881586.1

View Full Product Details