Product Details

SNP ID
rs116932132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76833109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTATTTGTAATAGTTTAGCTGT[A/G]GATTCTTTTGGAATTTTCTCTGTAA
Phenotype
MIM: 604760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF236 PubMed Links

Gene Details

Gene
ZNF236
Gene Name
zinc finger protein 236
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306089.1 Intron NP_001293018.1
NM_007345.3 Intron NP_031371.3
XM_005266762.3 Intron XP_005266819.1
XM_011526165.2 Intron XP_011524467.1
XM_011526166.2 Intron XP_011524468.1
XM_011526168.2 Intron XP_011524470.1
XM_011526169.2 Intron XP_011524471.1

View Full Product Details