Product Details
- SNP ID
-
rs116527960
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:79902982 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCCCACTCAGGAATTTGCTTCAG[C/G]CCAAAAGCCCAGGGCAGCAGGGTAA
- Phenotype
-
MIM: 605696
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs537273482] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNG2
- Gene Name
- potassium voltage-gated channel modifier subfamily G member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012283.1 |
1852 |
Intron |
|
|
NP_036415.1 |
- Gene
- PQLC1
- Gene Name
- PQ loop repeat containing 1
View Full Product Details