Product Details

SNP ID
rs115377178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57548962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATATCCTTAAGGGGAGAGAGT[A/G]GCTCCAAAAATAACCTTCCTTCCAG
Phenotype
MIM: 612386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FECH PubMed Links

Gene Details

Gene
FECH
Gene Name
ferrochelatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000140.3 3157 UTR 3 NP_000131.2
NM_001012515.2 3157 UTR 3 NP_001012533.1
XM_011525881.1 3157 UTR 3 XP_011524183.1
XM_011525882.2 3157 UTR 3 XP_011524184.1
XM_017025614.1 3157 Intron XP_016881103.1

View Full Product Details