Product Details

SNP ID
rs117354385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110817138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGATTGGTGCTAGGATAGGAC[A/G]TAACACATTCATTAGGAGGACAAAG
Phenotype
MIM: 180980 MIM: 614720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMD1 PubMed Links

Gene Details

Gene
AMD1
Gene Name
adenosylmethionine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287214.1 Intron NP_001274143.1
NM_001287215.1 Intron NP_001274144.1
NM_001287216.1 Intron NP_001274145.1
NM_001634.5 Intron NP_001625.2
Gene
CDK19
Gene Name
cyclin dependent kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300960.1 Intron NP_001287889.1
NM_001300963.1 Intron NP_001287892.1
NM_001300964.1 Intron NP_001287893.1
NM_015076.4 Intron NP_055891.1
XM_005266871.3 Intron XP_005266928.1
XM_011535630.2 Intron XP_011533932.1
XM_011535631.2 Intron XP_011533933.1
XM_011535632.1 Intron XP_011533934.1
XM_017010587.1 Intron XP_016866076.1
XM_017010588.1 Intron XP_016866077.1

View Full Product Details