Product Details

SNP ID
rs113246501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:115943090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAATGACCATACATCGGACTT[A/G]ATGCTGAATTTATTACTACGAATGG
Phenotype
MIM: 606573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 1873 Silent Mutation ATC,ATT I412I NP_002022.1
XM_005266880.4 1873 Silent Mutation ATC,ATT I412I XP_005266937.1
XM_005266881.2 1873 Silent Mutation ATC,ATT I412I XP_005266938.1
XM_005266882.4 1873 Silent Mutation ATC,ATT I412I XP_005266939.1
XM_011535653.2 1873 Silent Mutation ATC,ATT I412I XP_011533955.1
XM_011535654.2 1873 Silent Mutation ATC,ATT I412I XP_011533956.1
XM_011535655.2 1873 Silent Mutation ATC,ATT I412I XP_011533957.1
XM_011535656.2 1873 Silent Mutation ATC,ATT I299I XP_011533958.1
XM_017010645.1 1873 Silent Mutation ATC,ATT I412I XP_016866134.1

View Full Product Details