Product Details

SNP ID
rs116048760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118815808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTAGCCTTTTTTTCTTGTTACT[G/T]TCCACATTGTCTGCCCTCCATGGAA
Phenotype
MIM: 610098
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCM9 PubMed Links

Gene Details

Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 2463 Missense Mutation GAA,GAC E816D NP_060166.2
NM_153255.4 2463 Intron NP_694987.1

View Full Product Details